siRNA Details
| Gene Symbol: LINC01146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046094.1 | 4 | 563 | |||
| GenBank mRNA | BC029479.1 | - | 4 | 563 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG617119.1 | - | 4 | 563 | Chr. 14 - 88024550 - 88087259 [+] on Build GRCh38 | |

LINC01146
Genomic Map