siRNA Details
| Gene Symbol: LINC01164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038365.1 | 4 | 2030 | |||
| GenBank mRNA | AK128177.1 | - | 4 | 2030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01164| Gene Symbol: LINC01164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038365.1 | 4 | 2030 | |||
| GenBank mRNA | AK128177.1 | - | 4 | 2030 | ||