siRNA Details
| Gene Symbol: LOC101927809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134605.1 | 2 | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927809| Gene Symbol: LOC101927809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134605.1 | 2 | 319 | |||