siRNA Details
| Gene Symbol: TMEM108-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110812.1 | 3 | 1451 | |||
| GenBank mRNA | BC037929.2 | - | 3 | 1452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM108-AS1| Gene Symbol: TMEM108-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110812.1 | 3 | 1451 | |||
| GenBank mRNA | BC037929.2 | - | 3 | 1452 | ||