siRNA Details
Gene Symbol: LOC101929261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134277.1 | 2 | 805 | |||
GenBank mRNA | BC030121.2 | - | 2 | 805 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG700797.1 | - | 2 | 809 | Chr. 5 - 7347044 - 7347977 [-] on Build GRCh38 |
