siRNA Details
Gene Symbol: LOC101928461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125858.1 | 2 | 310 | |||
GenBank mRNA | BX093515.1 | - | 2 | 297 | ||
BX398118.2 | - | 2 | 317 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125858.1 | 2 | 310 | |||
GenBank mRNA | BX093515.1 | - | 2 | 297 | ||
BX398118.2 | - | 2 | 317 |