siRNA Details
| Gene Symbol: C5orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173667.3 | NP_775938.1 | 4 | 1586 | ||
| NR_126523.1 | 3 | 1398 | ||||
| XM_017009378.1 | XP_016864867.1 | 2 | 4005 | |||
| XM_017009379.1 | XP_016864868.1 | 2 | 4005 | |||
| XM_017009380.1 | XP_016864869.1 | 2 | 4005 | |||
| XM_017009381.1 | XP_016864870.1 | 2 | 4005 | |||
| XM_017009382.1 | XP_016864871.1 | 3 | 1507 | |||
| GenBank mRNA | AK094862.1 | - | 4 | 1557 | ||

C5orf64
Genomic Map