siRNA Details
| Gene Symbol: LOC100505817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038340.1 | 3 | 817 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG180747.1 | - | 2 | 565 | Chr. 18 - 73332463 - 73349736 [+] on Build GRCh38 | |

LOC100505817
Genomic Map