siRNA Details
| Gene Symbol: LOC100507389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038455.1 | 5 | 1495 | |||
| GenBank mRNA | AK096143.1 | - | 5 | 1496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507389| Gene Symbol: LOC100507389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038455.1 | 5 | 1495 | |||
| GenBank mRNA | AK096143.1 | - | 5 | 1496 | ||