siRNA Details
| Gene Symbol: CSMD2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038372.1 | 8 | 1079 | |||
| GenBank mRNA | BC031866.1 | - | 8 | 1093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSMD2-AS1| Gene Symbol: CSMD2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038372.1 | 8 | 1079 | |||
| GenBank mRNA | BC031866.1 | - | 8 | 1093 | ||