siRNA Details
| Gene Symbol: LOC105374266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135537.1 | 2 | 106 | |||
| NR_135538.1 | 2 | 106 | ||||
| GenBank mRNA | BX364319.2 | - | 2 | 106 | ||
| BX403058.2 | - | 2 | 106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374266| Gene Symbol: LOC105374266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135537.1 | 2 | 106 | |||
| NR_135538.1 | 2 | 106 | ||||
| GenBank mRNA | BX364319.2 | - | 2 | 106 | ||
| BX403058.2 | - | 2 | 106 | |||