siRNA Details
| Gene Symbol: LINCR-0002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX365943.2 | - | 1, 2 | 53 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINCR-0002| Gene Symbol: LINCR-0002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX365943.2 | - | 1, 2 | 53 | ||