siRNA Details
| Gene Symbol: LINC01580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120322.1 | 2 | 147 | |||
| NR_120323.1 | 2 | 147 | ||||
| GenBank mRNA | BC021741.1 | - | 2 | 152 | ||
| DB077575.1 | - | 2 | 147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01580| Gene Symbol: LINC01580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120322.1 | 2 | 147 | |||
| NR_120323.1 | 2 | 147 | ||||
| GenBank mRNA | BC021741.1 | - | 2 | 152 | ||
| DB077575.1 | - | 2 | 147 | |||