siRNA Details
| Gene Symbol: LOC101928134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120326.1 | 3 | 296 | |||
| NR_120327.1 | 3 | 282 | ||||
| GenBank mRNA | BX095733.1 | - | 3 | 296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928134| Gene Symbol: LOC101928134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120326.1 | 3 | 296 | |||
| NR_120327.1 | 3 | 282 | ||||
| GenBank mRNA | BX095733.1 | - | 3 | 296 | ||