siRNA Details
Gene Symbol: LOC100506470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109919.1 | 1 | 106 | |||
GenBank mRNA | BX106486.1 | - | 1 | 51 | ||
HY047124.1 | - | 1 | 106 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109919.1 | 1 | 106 | |||
GenBank mRNA | BX106486.1 | - | 1 | 51 | ||
HY047124.1 | - | 1 | 106 |