siRNA Details
| Gene Symbol: LOC105376382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134491.1 | 2 | 229 | |||
| GenBank mRNA | BF218358.1 | - | 2 | 230 | ||
| BG035368.1 | - | 206 | ||||
| CF130727.1 | - | 2 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376382| Gene Symbol: LOC105376382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134491.1 | 2 | 229 | |||
| GenBank mRNA | BF218358.1 | - | 2 | 230 | ||
| BG035368.1 | - | 206 | ||||
| CF130727.1 | - | 2 | 203 | |||