siRNA Details
| Gene Symbol: LOC105447648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131191.1 | 1 | 269 | |||
| GenBank mRNA | BX458411.2 | - | 1 | 269 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX445613.2 | - | 1 | 269 | Chr. 16 - 14363109 - 14370266 [-] on Build GRCh38 | |

LOC105447648
Genomic Map