siRNA Details
| Gene Symbol: CDKN2B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003529.3 | 18, 19 | 3252 | |||
| NR_047532.1 | 13, 14 | 2041 | ||||
| NR_047534.1 | 8, 9 | 1305 | ||||
| NR_047535.1 | 7, 8 | 1200 | ||||
| NR_047536.1 | 6, 7 | 1064 | ||||
| NR_047537.1 | 5, 6 | 1000 | ||||
| NR_047538.1 | 4, 5 | 864 | ||||
| NR_047543.1 | 8, 9 | 1334 | ||||
| GenBank mRNA | AB548314.1 | - | 13, 14 | 2041 | ||
| BC038540.1 | - | 2, 3 | 342 | |||
| DQ485453.1 | - | 18, 19 | 3251 | |||
| GQ495919.1 | - | 4, 5 | 753 | |||
| GQ495920.1 | - | 6, 7 | 953 | |||
| GQ495921.1 | - | 5, 6 | 889 | |||
| GQ495922.1 | - | 7, 8 | 1089 | |||
| GQ495923.1 | - | 8, 9 | 1223 | |||
| GQ495925.1 | - | 8, 9 | 1305 | |||

CDKN2B-AS1
Genomic Map