siRNA Details
| Gene Symbol: LOC440117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033970.1 | 3 | 637 | |||
| GenBank mRNA | BC037858.2 | - | 3 | 593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440117| Gene Symbol: LOC440117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033970.1 | 3 | 637 | |||
| GenBank mRNA | BC037858.2 | - | 3 | 593 | ||