siRNA Details
| Gene Symbol: LINC01224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126448.1 | 6 | 1450 | |||
| NR_126449.1 | 5 | 1276 | ||||
| GenBank mRNA | AK022793.1 | - | 5 | 1239 | ||
| BC041431.1 | - | 5 | 1282 | |||
| BC043213.1 | - | 8 | 1901 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01224| Gene Symbol: LINC01224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126448.1 | 6 | 1450 | |||
| NR_126449.1 | 5 | 1276 | ||||
| GenBank mRNA | AK022793.1 | - | 5 | 1239 | ||
| BC041431.1 | - | 5 | 1282 | |||
| BC043213.1 | - | 8 | 1901 | |||