siRNA Details
| Gene Symbol: LINC01224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126448.1 | 5 | 629 | |||
| NR_126449.1 | 4 | 455 | ||||
| GenBank mRNA | AK022793.1 | - | 4 | 418 | ||
| BC041431.1 | - | 4 | 461 | |||
| BC043213.1 | - | 6 | 744 | |||
| DA736731.1 | - | 5 | 593 | |||

LINC01224
Genomic Map