siRNA Details
| Gene Symbol: ARIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF099153.1 | - | 1 | 539 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY291621.1 | - | 1 | 251 | Chr. 3 - 48983729 - 48984173 [-] on Build GRCh38 | |

ARIH2
Genomic Map