siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028410.1 | 2, 3 | 235 | |||
| GenBank mRNA | BC144506.1 | - | 2, 3 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00207| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028410.1 | 2, 3 | 235 | |||
| GenBank mRNA | BC144506.1 | - | 2, 3 | 223 | ||