siRNA Details
Gene Symbol: LOC101929371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109861.1 | 2 | 390 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB299045.1 | - | 1 | 366 | Chr. 20 - 10006351 - 10006919 [+] on Build GRCh38 |
