siRNA Details
Gene Symbol: IL12A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108088.1 | 10 | 1767 | |||
GenBank mRNA | AK097161.1 | - | 11 | 1767 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IL12A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108088.1 | 10 | 1767 | |||
GenBank mRNA | AK097161.1 | - | 11 | 1767 |