siRNA Details
| Gene Symbol: LINC00692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034055.1 | 5 | 505 | |||
| GenBank mRNA | BC033370.1 | - | 5 | 508 | ||
| BC126324.1 | - | 4 | 342 | |||
| BC143987.1 | - | 4 | 345 | |||
| BC143998.1 | - | 4 | 342 | |||
| BC143999.1 | - | 4 | 345 | |||

LINC00692
Genomic Map