siRNA Details
| Gene Symbol: TMPO-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027157.1 | 2 | 1410 | |||
| GenBank mRNA | AK054586.1 | - | 1 | 42 | ||
| BC037346.2 | - | 1 | 751 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPO-AS1| Gene Symbol: TMPO-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027157.1 | 2 | 1410 | |||
| GenBank mRNA | AK054586.1 | - | 1 | 42 | ||
| BC037346.2 | - | 1 | 751 | |||