siRNA Details
| Gene Symbol: LINC01033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126379.1 | 1, 2 | 53 | |||
| GenBank mRNA | BG281020.1 | - | 1, 2 | 54 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG281099.1 | - | 1, 2 | 54 | Chr. 5 - 54320944 - 54415089 [+] on Build GRCh38 | |

LINC01033
Genomic Map