siRNA Details
| Gene Symbol: LOC101929565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125985.1 | 2, 3 | 118 | |||
| GenBank mRNA | BG196701.1 | - | 2, 3 | 154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929565| Gene Symbol: LOC101929565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125985.1 | 2, 3 | 118 | |||
| GenBank mRNA | BG196701.1 | - | 2, 3 | 154 | ||