siRNA Details
| Gene Symbol: LOC101929565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125985.1 | 3 | 220 | |||
| GenBank mRNA | BG196701.1 | - | 3 | 256 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG194680.1 | - | 3 | 219 | Chr. 1 - 212545695 - 212556065 [+] on Build GRCh38 | |

LOC101929565
Genomic Map