siRNA Details
| Gene Symbol: EVADR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125857.1 | 3 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EVADR| Gene Symbol: EVADR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125857.1 | 3 | 447 | |||