siRNA Details
| Gene Symbol: CASC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026939.1 | 5 | 2342 | |||
| NR_026940.1 | 4 | 2242 | ||||
| GenBank mRNA | BC036393.1 | - | 5 | 2343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC2| Gene Symbol: CASC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026939.1 | 5 | 2342 | |||
| NR_026940.1 | 4 | 2242 | ||||
| GenBank mRNA | BC036393.1 | - | 5 | 2343 | ||