siRNA Details
| Gene Symbol: LOC100505853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134646.1 | 2 | 1054 | |||
| GenBank mRNA | BC037384.1 | - | 2 | 1036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505853| Gene Symbol: LOC100505853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134646.1 | 2 | 1054 | |||
| GenBank mRNA | BC037384.1 | - | 2 | 1036 | ||