siRNA Details
| Gene Symbol: CHKB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021492.2 | 2 | 555 | |||
| GenBank mRNA | BJ989458.1 | - | 2 | 436 | ||
| CR980518.1 | - | 2 | 459 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY124358.1 | - | 2 | 458 | Chr. 22 - 50583124 - 50583885 [+] on Build GRCh38 | |

CHKB-AS1
Genomic Map