siRNA Details
| Gene Symbol: CHKB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021492.2 | 1, 2 | 301 | |||
| GenBank mRNA | BE393070.1 | - | 1, 2 | 200 | ||
| BJ989458.1 | - | 1, 2 | 182 | |||
| CR456603.1 | - | 1, 2 | 301 | |||
| CR980518.1 | - | 1, 2 | 205 | |||

CHKB-AS1
Genomic Map