siRNA Details
| Gene Symbol: SNHG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003239.1 | 5 | 1016 | |||
| GenBank mRNA | AK055434.1 | - | 3 | 2582 | ||
| BC040237.1 | - | 3 | 2464 | |||
| Gene Symbol: SNORA60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091209.1 | - | 1 | 3015 | ||

SNHG11
Genomic Map