siRNA Details
| Gene Symbol: LOC102723377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120672.1 | 3 | 354 | |||
| GenBank mRNA | BX103495.1 | - | 3 | 355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723377| Gene Symbol: LOC102723377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120672.1 | 3 | 354 | |||
| GenBank mRNA | BX103495.1 | - | 3 | 355 | ||