siRNA Details
| Gene Symbol: LOC101928333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110194.1 | 3 | 1544 | |||
| NR_110195.1 | 2 | 1308 | ||||
| GenBank mRNA | BC040328.1 | - | 3 | 1543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928333| Gene Symbol: LOC101928333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110194.1 | 3 | 1544 | |||
| NR_110195.1 | 2 | 1308 | ||||
| GenBank mRNA | BC040328.1 | - | 3 | 1543 | ||