siRNA Details
| Gene Symbol: NCK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110175.1 | 1 | 78 | |||
| NR_110176.1 | 1 | 78 | ||||
| NR_110177.1 | 1 | 78 | ||||
| GenBank mRNA | CB047433.1 | - | 1 | 78 | ||
| DA714434.1 | - | 1 | 46 | |||
| HY007950.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCK1-AS1| Gene Symbol: NCK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110175.1 | 1 | 78 | |||
| NR_110176.1 | 1 | 78 | ||||
| NR_110177.1 | 1 | 78 | ||||
| GenBank mRNA | CB047433.1 | - | 1 | 78 | ||
| DA714434.1 | - | 1 | 46 | |||
| HY007950.1 | - | 1 | 78 | |||