siRNA Details
| Gene Symbol: ZNF790-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040027.1 | 6 | 722 | |||
| NR_040028.1 | 4 | 528 | ||||
| GenBank mRNA | BM467896.1 | - | 4 | 316 | ||
| BM476885.1 | - | 6 | 722 | |||
| BX394102.2 | - | 4 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF790-AS1| Gene Symbol: ZNF790-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040027.1 | 6 | 722 | |||
| NR_040028.1 | 4 | 528 | ||||
| GenBank mRNA | BM467896.1 | - | 4 | 316 | ||
| BM476885.1 | - | 6 | 722 | |||
| BX394102.2 | - | 4 | 527 | |||