siRNA Details
Gene Symbol: ZNF790-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040027.1 | 6 | 867 | |||
NR_040028.1 | 4 | 673 | ||||
GenBank mRNA | BM467896.1 | - | 4 | 461 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF790-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040027.1 | 6 | 867 | |||
NR_040028.1 | 4 | 673 | ||||
GenBank mRNA | BM467896.1 | - | 4 | 461 |