siRNA Details
| Gene Symbol: MIR325HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110400.2 | 4 | 1043 | |||
| NR_110401.2 | 4 | 1027 | ||||
| NR_110402.2 | 4 | 981 | ||||
| NR_110403.2 | 4 | 959 | ||||
| NR_110404.2 | 3 | 922 | ||||
| NR_110405.2 | 3 | 865 | ||||
| NR_110406.2 | 3 | 860 | ||||
| GenBank mRNA | AK057746.1 | - | 2 | 483 | ||
| AK311342.1 | - | 4 | 981 | |||

MIR325HG
Genomic Map