siRNA Details
| Gene Symbol: BRCAT54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109862.1 | 4 | 588 | |||
| NR_109863.1 | 4 | 584 | ||||
| NR_109864.1 | 3 | 523 | ||||
| NR_109865.1 | 3 | 519 | ||||
| GenBank mRNA | AL691871.1 | - | 3 | 508 | ||
| BG612611.1 | - | 4 | 525 | |||
| BY797234.2 | - | 4 | 533 | |||
| HY123663.1 | - | 3 | 511 | |||

BRCAT54
Genomic Map