siRNA Details
Gene Symbol: BRCAT54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109862.1 | 4 | 362 | |||
NR_109863.1 | 4 | 358 | ||||
NR_109864.1 | 3 | 297 | ||||
NR_109865.1 | 3 | 293 | ||||
GenBank mRNA | AL691871.1 | - | 3 | 282 | ||
BG612611.1 | - | 4 | 299 | |||
BY797234.2 | - | 4 | 307 | |||
HY123663.1 | - | 3 | 285 |
