siRNA Details
Gene Symbol: VLDLR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015375.2 | 2 | 415 | |||
GenBank mRNA | AF424541.1 | - | 6 | 880 | ||
BC004474.2 | - | 2 | 415 | |||
DA119626.1 | - | 2 | 352 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VLDLR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015375.2 | 2 | 415 | |||
GenBank mRNA | AF424541.1 | - | 6 | 880 | ||
BC004474.2 | - | 2 | 415 | |||
DA119626.1 | - | 2 | 352 |