siRNA Details
Gene Symbol: VLDLR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015375.2 | 2 | 317 | |||
GenBank mRNA | AF424541.1 | - | 6 | 782 | ||
BC004474.2 | - | 2 | 317 | |||
BE086096.1 | - | 2 | 125 | |||
DA119626.1 | - | 2 | 254 |
