siRNA Details
| Gene Symbol: LINC00868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135283.1 | 1 | 15 | |||
| NR_135284.1 | 1 | 15 | ||||
| GenBank mRNA | BX089989.1 | - | 1 | 17 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG189496.1 | - | 1 | 16 | Chr. 17 - 76856168 - 76861838 [+] on Build GRCh38 | |

LINC00868
Genomic Map