siRNA Details
| Gene Symbol: DIAPH2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125391.1 | 2 | 191 | |||
| GenBank mRNA | AB593036.1 | - | 2 | 191 | ||
| AB593037.1 | - | 2 | 189 | |||
| L22650.1 | - | 3 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIAPH2-AS1| Gene Symbol: DIAPH2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125391.1 | 2 | 191 | |||
| GenBank mRNA | AB593036.1 | - | 2 | 191 | ||
| AB593037.1 | - | 2 | 189 | |||
| L22650.1 | - | 3 | 439 | |||