siRNA Details
| Gene Symbol: MIR99AHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136544.1 | 1 | 466 | |||
| NR_136545.1 | 1 | 466 | ||||
| NR_136546.1 | 1 | 466 | ||||
| GenBank mRNA | DA338933.1 | - | 1 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR99AHG| Gene Symbol: MIR99AHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136544.1 | 1 | 466 | |||
| NR_136545.1 | 1 | 466 | ||||
| NR_136546.1 | 1 | 466 | ||||
| GenBank mRNA | DA338933.1 | - | 1 | 299 | ||