siRNA Details
| Gene Symbol: NPSR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033664.1 | 1, 2 | 205 | |||
| NR_033665.1 | 1, 2 | 205 | ||||
| GenBank mRNA | AY312367.1 | - | LIT3327 AAA1 |
1, 2 | 205 | |
| AY312368.1 | - | LIT3328 AAA1 |
1, 2 | 205 | ||
| AY312369.1 | - | LIT3332 AAA1 |
1, 2 | 205 | ||
| AY312371.1 | - | LIT3334 AAA1 |
1, 2 | 205 | ||
| AY312372.1 | - | LIT3335 AAA1 |
1, 2 | 205 | ||
| BX649082.1 | - | 1, 2 | 168 | |||
| U85653.1 | - | 1, 2 | 168 | |||

NPSR1-AS1
Genomic Map